
Benzamidine hydrochloride hydrate

Synonym(s):
Amidinobenzene hydrochloride, Benzamidinium chloride, Benzenecarboximidamide hydrochloride
Linear Formula:
C6H5C(=NH)NH2 · HCl · xH2O
Products
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing