
Benserazide impurity A

Synonym(s):
DL-Serine hydrazide, (RS)-2-Amino-3-hydroxypropanohydrazide
Empirical Formula (Hill Notation):
C3H9N3O2
CAS Number:
Molecular Weight:
119.12
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing