Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Barium
Empirical Formula (Hill Notation):
Ba
CAS Number:
7440-39-3
Molecular Weight:
137.33
Products
Brand
Sigma-Aldrich (6)
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
474711
Barium,
dendritic pieces, purified by distillation, 99.99% trace metals basis
Expand
View Pricing
441880
Barium,
dendritic pieces, purified by distillation, 99.9% trace metals basis
Expand
View Pricing
735787
Barium,
beads, 0.5-2.0 mm particle size, 99% trace metals basis
Expand
View Pricing
403334
Barium,
pieces, 1 cm, 99% trace metals basis
Expand
View Pricing
237094
Barium,
rod, diam. ~2 cm, ≥99% trace metals basis
Expand
View Pricing
938386
Barium,
beads, ≤2.0 mm particle size, 99.99% trace metals basis
Expand
View Pricing