
Azure B

Synonym(s):
N,N,N′-Trimethylthionin, Azure I, Methyleneazure
Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Chemical Composition
Quality Segment
Application
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing