Skip to Content
Merck
Aztreonam analytical standard

Aztreonam

Aztreonam analytical standard
Synonym(s):
[2S-[2α,3β(Z)]]-2-[[[1-(2-Amino-4-thiazolyl)-2-[(2-methyl-4-oxo-1-sulfo-3-azetidinyl)amino]-2-oxoethylidene]amino]oxy]-2-methylpropanoic acid, Aztreonam
Empirical Formula (Hill Notation):
C13H17N5O8S2
CAS Number:
Molecular Weight:
435.43
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique