
Arg-Gly-Asp-Ser

Empirical Formula (Hill Notation):
C15H27N7O8
CAS Number:
Molecular Weight:
433.42
Products
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing