
Arachidonylethanolamide

Synonym(s):
AEA, Anandamide, Arachidonic acid N-(hydroxyethyl)amide
Empirical Formula (Hill Notation):
C22H37NO2
CAS Number:
Molecular Weight:
347.53
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing