Anti-Seasonal H1N1 Hemagglutinin antibody produced in rabbit
Synonym(s):
Anti-Common flu H1, Anti-Flu H1, Anti-HA, Anti-Seasonal Influenza A (H1N1) Hemagglutinin
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2