
Antipyrine

Synonym(s):
2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone, Antipyrine
Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3