
Anti-B4GALT1 antibody produced in rabbit

Synonym(s):
Anti-β-1,4-GalTase 1 antibody produced in rabbit, Anti-β-1,4-Galactosyltransferase 1 antibody produced in rabbit, Anti-β4Gal-T1 antibody produced in rabbit, Anti-UDP-Gal:β-GlcNAc β-1,4-galactosyltransferase 1 antibody produced in rabbit, Anti-UDP-galactose:β-N-acetylglucosamine β-1,4-galactosyltransferase 1 antibody produced in rabbit, Anti-b4Gal-T1 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1