
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-Actin, α-Smooth Muscle, SMA
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing