Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Angled Tips
Products
Brand
Sigma-Aldrich (21)
Color
Length (in)
Package Size
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
931276
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 14 gauge × 0.5 in. × 0.060 in., stainless steel tip, olive green, box of 50 pieces
Expand
View Pricing
931292
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 14 gauge × 0.5 in. × 0.060 in., stainless steel tip, olive green, box of 50 pieces
Expand
View Pricing
931357
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 20 gauge × 0.5 in. × 0.024 in., stainless steel tip, pink, box of 50 pieces
Expand
View Pricing
931330
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 18 gauge × 0.5 in. × 0.033 in., stainless steel tip, green, box of 50 pieces
Expand
View Pricing
931489
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 30 gauge × 0.5 in. × 0.006 in., stainless steel tip, lavender, box of 50 pieces
Expand
View Pricing
931497
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 30 gauge × 0.5 in. × 0.006 in., stainless steel tip, lavender, box of 50 pieces
Expand
View Pricing
931411
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 23 gauge × 0.5 in. × 0.013 in., stainless steel tip, orange, box of 50 pieces
Expand
View Pricing
931349
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 20 gauge × 0.5 in. × 0.024 in., stainless steel tip, pink, box of 50 pieces
Expand
View Pricing
931381
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 22 gauge × 0.5 in. × 0.016 in., stainless steel tip, blue, box of 50 pieces
Expand
View Pricing
931438
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 23 gauge × 0.5 in. × 0.013 in., stainless steel tip, orange, box of 50 pieces
Expand
View Pricing
931462
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 27 gauge × 0.5 in. × 0.008 in., stainless steel tip, clear, box of 50 pieces
Expand
View Pricing
931454
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 25 gauge × 0.5 in. × 0.010 in., stainless steel tip, red, box of 50 pieces
Expand
View Pricing
931284
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 14 gauge × 1.5 in. × 0.060 in., stainless steel tip, olive green, box of 50 pieces
Expand
View Pricing
931373
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 21 gauge × 1.5 in. × 0.02 in., stainless steel tip, purple, box of 50 pieces
Expand
View Pricing
931403
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 22 gauge × 0.5 in. × 0.016 in., stainless steel tip, blue, box of 50 pieces
Expand
View Pricing
931306
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 15 gauge × 0.5 in. × 0.053 in., stainless steel tip, amber, box of 50 pieces
Expand
View Pricing
931446
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 25 gauge × 0.5 in. × 0.010 in., stainless steel tip, red, box of 50 pieces
Expand
View Pricing
931470
Angled Tips,
angle, 90 ° bend, tip size × L × I.D. 27 gauge × 0.5 in. × 0.008 in., stainless steel tip, clear, box of 50 pieces
Expand
View Pricing
931365
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 21 gauge × 0.5 in. × 0.020 in., stainless steel tip, purple, box of 50 pieces
Expand
View Pricing
931322
Angled Tips,
angle, 45 ° bend, tip size × L × I.D. 18 gauge × 1.5 in. × 0.033 in., stainless steel tip, green, box of 50 pieces
Expand
View Pricing
Page 1 of 2
Page 1 of 2