Andrographolide
Synonym(s):
Andrographolide, (3E,4S)-3-[2-[(1R,4aS,5R,6R,8aS)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethylidene]dihydro-4-hydroxy-2(3H)-furanone, ent-(3β,12E,14R)-3,14,19-Trihydroxy-8(17),12-labdadien-16,15-olide, Andrographis
Empirical Formula (Hill Notation):
C20H30O5
CAS Number:
Molecular Weight:
350.45
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing