Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Amodiaquin dihydrochloride dihydrate
Synonym(s):
4-[(7-Chloro-4-quinolinyl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride dihydrate, 4-([7-Chloro-4-quinolinyl]amino)-2-([diethylamino]methyl)phenol
Empirical Formula (Hill Notation):
C
20
H
22
ClN
3
O · 2HCl · 2H
2
O
CAS Number:
6398-98-7
Molecular Weight:
464.81
EC Number:
200-706-0
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
CDS022953
Amodiaquin dihydrochloride dihydrate,
Expand
View Pricing
A2799
Amodiaquin dihydrochloride dihydrate,
analytical standard
Expand
View Pricing