Amicarbazone
Synonym(s):
4-Amino-N-tert-butyl-4,5-dihydro-3-isopropyl-5-oxo-1,2,4-1H-triazole-1-carboxamide
Empirical Formula (Hill Notation):
C10H19N5O2
CAS Number:
Molecular Weight:
241.29
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1