Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum/Silicon
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF13750006
Aluminum/Silicon,
foil, Al 99%/Si 1%, thickness 0.075 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF93359560
Aluminum/Silicon,
foil, thickness 0.075 mm, Al99%/Si 1%, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF05754359
Aluminum/Silicon,
rod, Al 88%/Si 12%, 10 mm diameter, length 100 mm, condition extruded
Expand
View Pricing
GF69566620
Aluminum/Silicon,
rod, Al99%/Si 1%, 5 mm diameter, bulk density 2.7 g/cm
3
, length 100 mm
Expand
View Pricing
GF97477611
Aluminum/Silicon,
rod, 5 mm diameter, Al99%/Si 1%, length 200 mm, bulk density 2.7 g/cm
3
Expand
View Pricing
GF94319095
Aluminum/Silicon,
foil, thickness 0.075 mm, Al99%/Si 1%, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF37581699
Aluminum/Silicon,
rod, Al 99%/Si 1%, 5 mm diameter, length 1000 mm, bulk density 2.7 g/cm
3
Expand
View Pricing
GF94463725
Aluminum/Silicon,
foil, thickness 0.075 mm, Al99%/Si 1%, size 150 × 150 mm, temper as rolled
Expand
View Pricing
GF75012724
Aluminum/Silicon,
rod, Al88%/Si12%, 10 mm diameter, length 1000 mm, condition extruded
Expand
View Pricing
GF48132648
Aluminum/Silicon,
rod, Al 99%/Si 1%, 5 mm diameter, length 500 mm, bulk density 2.7 g/cm
3
Expand
View Pricing