Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum alloy 5251
Products
Manufacturer
Physical Form
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF81042252
Aluminum alloy 5251,
foil, Al97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 250 × 250 mm, condition mirror polished on both sides
Expand
View Pricing
GF32180149
Aluminum alloy 5251,
foil, Al 97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 100 × 100 mm, condition mirror polished on both sides
Expand
View Pricing
GF10241413
Aluminum alloy 5251,
foil, Al 97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 150 × 150 mm, condition mirror polished on both sides
Expand
View Pricing
GF12573918
Aluminum alloy 5251,
foil, Al 97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 150 × 150 mm
Expand
View Pricing
GF18966190
Aluminum alloy 5251,
foil, Al 97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 50 × 50 mm
Expand
View Pricing
GF15702454
Aluminum alloy 5251,
foil, Al 97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 100 × 100 mm
Expand
View Pricing
GF84620437
Aluminum alloy 5251,
foil, thickness 1.5 mm, Al97.7%/Mg 2%/Mn 0.3%, size 50 × 50 mm, condition mirror polished on both sides
Expand
View Pricing
GF05868311
Aluminum alloy 5251,
foil, Al 97.7%/Mg 2%/Mn 0.3%, thickness 1.5 mm, size 500 × 500 mm, condition mirror polished on both sides
Expand
View Pricing