Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminium alloy 2014
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF06811107
Aluminium alloy 2014,
rod, Al 92%/Cu 8%/Mn 1%/Si 1%/Fe 0.5%/Mg, 22.2 mm diameter, length 1000 mm
Expand
View Pricing
GF13414586
Aluminium alloy 2014,
rod, Al 92%/Cu 8%/Mn 1%/Si 1%/Fe 0.5%/Mg, 22.2 mm diameter, length 100 mm
Expand
View Pricing
GF36029493
Aluminium alloy 2014,
rod, Al 92%/Cu 8%/Mn 1%/Si 1%/Fe 0.5%/Mg, 22.2 mm diameter, length 500 mm
Expand
View Pricing
GF90000683
Aluminium alloy 2014,
rod, Al92%/Cu 8%/Mn 1%/Si 1%/Fe 0.5%/Mg %, length 200 mm, 22.2 mm diameter
Expand
View Pricing
GF75163833
Aluminium alloy 2014,
foil, Al92%/Cu 8%/Mn 1%/Si 1%/Fe 0.5%/Mg %, thickness 1.50 mm, size 150 × 150 mm
Expand
View Pricing