Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Actinomycin D
Synonym(s):
Actinomycin C
1
, Actinomycin IV, Dactinomycin
Empirical Formula (Hill Notation):
C
62
H
86
N
12
O
16
CAS Number:
50-76-0
Molecular Weight:
1255.42
EC Number:
200-063-6
Products
Product Category
antibiotics (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A1410
Actinomycin D,
from
Streptomyces
sp., ~98% (HPLC)
Expand
View Pricing
A9415
Actinomycin D,
from
Streptomyces
sp., suitable for cell culture, ≥95%
Expand
View Pricing
A4262
Actinomycin D,
from
Streptomyces
sp., ≥95% (HPLC)
Expand
View Pricing