Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace pressure tube
Products
Product Category
tubes (33)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Length (cm)
OD (mm)
Color
Sterility
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z181099
Ace pressure tube,
bushing type, Front seal, capacity ~15 mL, L × O.D. 10.2 cm × 25.4 mm
Expand
View Pricing
Z566268
Ace pressure tube,
Front seal, capacity 120 mL, 150 PSI, #15 plug, L × O.D. 17.8 cm × 38.1 mm
Expand
View Pricing
Z181064
Ace pressure tube,
bushing type, Back seal, capacity ~15 mL, L × O.D. 10.2 cm × 25.4 mm
Expand
View Pricing
Z181110
Ace pressure tube,
bushing type, Front seal, capacity ~38 mL, L × O.D. 20.3 cm × 25.4 mm
Expand
View Pricing
Z568821
Ace pressure tube,
bushing type, Front seal, capacity 140 mL, L × O.D. 20.3 cm × 38.1 mm
Expand
View Pricing
Z566241
Ace pressure tube,
bushing type, Back seal, capacity 100 mL, L × O.D. 17.8 cm × 38.1 mm
Expand
View Pricing
Z564567
Ace pressure tube,
bushing type, Front seal, capacity ~9 mL, L × O.D. 10.2 cm × 19 mm
Expand
View Pricing
Z564559
Ace pressure tube,
bushing type, Front seal, capacity ~4 mL, L × O.D. 10.2 cm × 13 mm
Expand
View Pricing
Z566276
Ace pressure tube,
bushing type, Back seal, capacity 200 mL, L × O.D. 20.3 cm × 50 mm
Expand
View Pricing
Z564583
Ace pressure tube,
bushing type, Front seal, capacity ~8 mL, L × O.D. 17.8 cm × 13 mm
Expand
View Pricing
Z564540
Ace pressure tube,
bushing type, Front seal, capacity ~1 mL, L × O.D. 10.2 cm × 8 mm
Expand
View Pricing
Z181080
Ace pressure tube,
bushing type, Back seal, capacity ~38 mL, L × O.D. 20.3 cm × 25.4 mm
Expand
View Pricing
Z568805
Ace pressure tube,
bushing type, Front seal, capacity 60 mL, L × O.D. 10.2 cm × 38.1 mm
Expand
View Pricing
Z564621
Ace pressure tube,
bushing type, Front seal, capacity ~21 mL, L × O.D. 20.3 cm × 19 mm
Expand
View Pricing
Z564575
Ace pressure tube,
bushing type, Front seal, capacity ~2 mL, L × O.D. 17.8 cm × 8 mm
Expand
View Pricing
Z568767
Ace pressure tube,
bushing type, Front seal, capacity 60 mL, L × O.D. 10.2 cm × 38.1 mm
Expand
View Pricing
Z181102
Ace pressure tube,
bushing type, Front seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Expand
View Pricing
Z568856
Ace pressure tube,
bushing type, Back seal, capacity 48 mL, L × O.D. 10.2 cm × 38.1 mm
Expand
View Pricing
Z181072
Ace pressure tube,
bushing type, Back seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Expand
View Pricing
Z568910
Ace pressure tube,
bushing type, Back seal, capacity 140 mL, L × O.D. 20.3 cm × 38.1 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2