Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace evaporating flask, pear-shaped
Synonym(s):
Aldrich
®
evaporating flask, pear-shaped
Products
Product Category
flasks (10)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z682535
Ace evaporating flask, pear-shaped,
capacity 1000 mL, joint: ST/NS 29/42, non-coated, heavy wall
Expand
View Pricing
Z682594
Ace evaporating flask, pear-shaped,
capacity 1000 mL, joint: ST/NS 24/40, poly-coated, heavy wall
Expand
View Pricing
Z682578
Ace evaporating flask, pear-shaped,
capacity 500 mL, joint: ST/NS 29/32, poly-coated, heavy wall
Expand
View Pricing
Z682616
Ace evaporating flask, pear-shaped,
capacity 1000 mL, joint: ST/NS 29/42, poly-coated, heavy wall
Expand
View Pricing
Z682527
Ace evaporating flask, pear-shaped,
capacity 500 mL, joint: ST/NS 29/42, non-coated, heavy wall
Expand
View Pricing
Z682608
Ace evaporating flask, pear-shaped,
capacity 1000 mL, joint: ST/NS 29/32, poly-coated, heavy wall
Expand
View Pricing
Z682632
Ace evaporating flask, pear-shaped,
capacity 2000 mL, joint: ST/NS 29/32, poly-coated, heavy wall
Expand
View Pricing
Z682624
Ace evaporating flask, pear-shaped,
capacity 2000 mL, joint: ST/NS 24/40, poly-coated, heavy wall
Expand
View Pricing
Z682551
Ace evaporating flask, pear-shaped,
capacity 500 mL, joint: ST/NS 24/40, poly-coated, heavy wall
Expand
View Pricing
Z682640
Ace evaporating flask, pear-shaped,
capacity 2000 mL, joint: ST/NS 29/42, poly-coated, heavy wall
Expand
View Pricing