Skip to Content
Merck
α-Bromoisobutyryl bromide 98%

α-Bromoisobutyryl bromide

α-Bromoisobutyryl bromide 98%
Synonym(s):
2-Bromo-2-methylpropionyl bromide, BIBB
Linear Formula:
(CH3)2CBrCOBr
CAS Number:
Molecular Weight:
229.90
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing