Skip to Content
Merck
7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid 96%

7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid

7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid 96%
Synonym(s):
Fluoroquinolonic acid, Q-Acid
Empirical Formula (Hill Notation):
C13H9ClFNO3
CAS Number:
Molecular Weight:
281.67
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1