![5-Cholesten-3β-ol-7-one ≥90%](/deepweb/assets/sigmaaldrich/product/structures/129/728/abf92da5-dfb8-4ba6-9ae5-291d1a2aa834/640/abf92da5-dfb8-4ba6-9ae5-291d1a2aa834.png)
5-Cholesten-3β-ol-7-one
![5-Cholesten-3β-ol-7-one ≥90%](/deepweb/assets/sigmaaldrich/product/structures/129/728/abf92da5-dfb8-4ba6-9ae5-291d1a2aa834/640/abf92da5-dfb8-4ba6-9ae5-291d1a2aa834.png)
Synonym(s):
3β-Hydroxy-5-cholesten-7-one, 7-Ketocholesterol
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing