Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside
Synonym(s):
5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal, 5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside
Empirical Formula (Hill Notation):
C
14
H
15
BrClNO
6
CAS Number:
7240-90-6
Molecular Weight:
408.63
EC Number:
230-640-8
Products
Product Category
substrates (4)
bioactive small molecules (1)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
B9146
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside,
≥98%, powder, suitable for identification of lac+ bacterial colonies
Expand
View Pricing
B4252
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside,
chromogenic, ≥98%, powder
Expand
View Pricing
203782
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside,
A chromogenic substrate for β-galactosidase. Used to distinguish between recombinant and non-recombinant plasmids carrying β-galactosidase gene (e.g. pUR222).
Expand
View Pricing
B6024
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside,
tablet, suitable for identification of lac+ bacterial colonies
Expand
View Pricing