Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Azacytosine
Synonym(s):
2-Hydroxy-4-aminotriazine, 4-Amino-1,3,5-triazin-2-ol, 4-Amino-1,3,5-triazin-2-one, 4-Amino-2-hydroxy-1,3,5-triazine, NSC 51100, NSC 54006
Empirical Formula (Hill Notation):
C
3
H
4
N
4
O
CAS Number:
931-86-2
Molecular Weight:
112.09
EC Number:
213-242-9
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
855049
5-Azacytosine,
Expand
View Pricing