
5,10,15,20-Tetraphenyl-21H,23H-porphine manganese(III) chloride

Synonym(s):
meso-Tetraphenylporphyrin manganese(III) chloride complex
Empirical Formula (Hill Notation):
C44H28ClMnN4
CAS Number:
Molecular Weight:
703.11
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing