Skip to Content
Merck
4-tert-Butylcyclohexyl acetate, mixture of cis and trans 99%

4-tert-Butylcyclohexyl acetate, mixture of cis and trans

4-tert-Butylcyclohexyl acetate, mixture of cis and trans 99%
Linear Formula:
CH3CO2C6H10C(CH3)3
CAS Number:
Molecular Weight:
198.30
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing