Skip to Content
Merck
4-Oxoisophorone ≥98%, FG

4-Oxoisophorone

4-Oxoisophorone ≥98%, FG
Synonym(s):
2,6,6-Trimethyl-2-cyclohexene-1,4-dione, 4-Oxoisophorone, Ketoisophorone
Empirical Formula (Hill Notation):
C9H12O2
CAS Number:
Molecular Weight:
152.19
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing