Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4,
N
,
N
-Trimethylaniline
Synonym(s):
4-Dimethylaminotoluene,
N
,
N
-Dimethyl-
p
-toluidine
Linear Formula:
CH
3
C
6
H
4
N(CH
3
)
2
CAS Number:
99-97-8
Molecular Weight:
135.21
EC Number:
202-805-4
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Puriss (1)
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D189006
4,
N
,
N
-Trimethylaniline,
99%
Expand
View Pricing
41690
4,
N
,
N
-Trimethylaniline,
catalyst grade (for peroxide polymerization), ≥98.5% (GC)
Expand
View Pricing
41700
4,
N
,
N
-Trimethylaniline,
purum, ≥98.0% (GC)
Expand
View Pricing