Skip to Content
Merck
4-Imidazolemethanol hydrochloride 98%

4-Imidazolemethanol hydrochloride

4-Imidazolemethanol hydrochloride 98%
Synonym(s):
4-(Hydroxymethyl)imidazole hydrochloride
Empirical Formula (Hill Notation):
C4H6N2O · HCl
CAS Number:
Molecular Weight:
134.56
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing