Skip to Content
Merck
4-Hydroxybenzoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

4-Hydroxybenzoic acid

4-Hydroxybenzoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
4-Hydroxybenzoic acid
Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique