
4-Fluorophenylmagnesium bromide solution

Linear Formula:
FC6H4MgBr
CAS Number:
Molecular Weight:
199.30
Products
Brand
Formula Weight
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing