Skip to Content
Merck
4-Chlorobenzaldehyde 97%

4-Chlorobenzaldehyde

4-Chlorobenzaldehyde 97%
Synonym(s):
4-Chlorobenzaldehyde
Linear Formula:
ClC6H4CHO
CAS Number:
Molecular Weight:
140.57

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing