Skip to Content
Merck
4-(Bromomethyl)benzonitrile 99%

4-(Bromomethyl)benzonitrile

4-(Bromomethyl)benzonitrile 99%
Synonym(s):
α-Bromo-p-tolunitrile, 4-Cyanobenzyl bromide
Linear Formula:
BrCH2C6H4CN
CAS Number:
Molecular Weight:
196.04
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing