
4-(Benzyloxy)benzyl bromide, polymer-bound

Synonym(s):
4-(Bromomethyl)phenoxymethylpolystyrene, Wang brominated resin
Products
Brand
Functional Group
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing