Skip to Content
Merck
4-Arm PEG10K - Dibenzocyclooctyne

4arm-PEG10K

4-Arm PEG10K - Dibenzocyclooctyne
Synonym(s):
4arm-PEG-Succinimidyl Carboxymethyl Ester, 4arm-PEG-Maleimide, 4arm-PEG-SH, 4arm-PEG-NH2, 4arm-PEG-COOH
Linear Formula:
C(CH2O(CH2CH2O)nCH2CH2NCO)4