Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Synonym(s):
1-Amino-2-naphthol-4-sulfonic acid
Linear Formula:
H
2
NC
10
H
5
(OH)SO
3
H
CAS Number:
116-63-2
Molecular Weight:
239.25
EC Number:
204-147-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Special Grade
ACS reagent (1)
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
398969
4-Amino-3-hydroxy-1-naphthalenesulfonic acid,
ACS reagent, ≥90%
Expand
View Pricing
08751
4-Amino-3-hydroxy-1-naphthalenesulfonic acid,
for spectrophotometric det. of Si, ≥90.0% (CHN)
Expand
View Pricing