
4,4′-DDT-d8

Synonym(s):
1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane-d8
Empirical Formula (Hill Notation):
C14HCl5D8
Molecular Weight:
362.54
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing