
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl

Synonym(s):
4,4′-Bis(9-carbazolyl)-1,1′-biphenyl, 4,4-N,N′-Dicarbazole-1,1′-biphenyl, CBP, DCBP
Empirical Formula (Hill Notation):
C36H24N2
CAS Number:
Molecular Weight:
484.59
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing