Skip to Content
Merck
4,4′-(1,3-Phenylenediisopropylidene)bisphenol 99%

4,4′-(1,3-Phenylenediisopropylidene)bisphenol

4,4′-(1,3-Phenylenediisopropylidene)bisphenol 99%
Synonym(s):
Bisphenol M
Linear Formula:
C6H4[C(CH3)2C6H4OH]2
CAS Number:
Molecular Weight:
346.46

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing