Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Pentanone
Synonym(s):
Diethyl ketone
Linear Formula:
CH
3
CH
2
COCH
2
CH
3
CAS Number:
96-22-0
Molecular Weight:
86.13
EC Number:
202-490-3
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (2)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Organoleptic Class
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W510106
3-Pentanone,
≥99%
Expand
View Pricing
127604
3-Pentanone,
ReagentPlus
®
, ≥99.0% (GC)
Expand
View Pricing