Skip to Content
Merck
3-Mercaptohexyl acetate ≥98%, FG

3-Mercaptohexyl acetate

3-Mercaptohexyl acetate ≥98%, FG
Empirical Formula (Hill Notation):
C8H16O2S
CAS Number:
Molecular Weight:
176.28

Products

Product Category
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing