
3-Fluorophenylmagnesium bromide solution

Linear Formula:
FC6H4MgBr
CAS Number:
Molecular Weight:
199.30
Products
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing