Skip to Content
Merck
3-Chloroperbenzoic acid ≤77%

3-Chloroperbenzoic acid

3-Chloroperbenzoic acid ≤77%
Synonym(s):
3-Chloroperoxybenzoic acid, MCPBA
Linear Formula:
ClC6H4CO3H
CAS Number:
Molecular Weight:
172.57

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing