Skip to Content
Merck
3-(Chloromethyl)benzoic acid 97%

3-(Chloromethyl)benzoic acid

3-(Chloromethyl)benzoic acid 97%
Synonym(s):
α-Chloro-m-toluic acid
Linear Formula:
ClCH2C6H4CO2H
CAS Number:
Molecular Weight:
170.59

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing