Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Chloro-2-methyl-1-propene
Synonym(s):
β-Methallyl chloride, 3-Chloro-2-methyl-1-propene
Linear Formula:
CH
2
=C(CH
3
)CH
2
Cl
CAS Number:
563-47-3
Molecular Weight:
90.55
EC Number:
209-251-2
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
108030
3-Chloro-2-methyl-1-propene,
technical grade, 90%
Expand
View Pricing
8.05905
3-Chloro-2-methyl-1-propene,
for synthesis
Expand
View Pricing
281972
3-Chloro-2-methyl-1-propene,
contains ~250 ppm Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate) as stabilizer, 98%
Expand
View Pricing