Skip to Content
Merck
(3-Aminopropyl)trimethoxysilane 97%

(3-Aminopropyl)trimethoxysilane

(3-Aminopropyl)trimethoxysilane 97%
Synonym(s):
3-(Trimethoxysilyl)propylamine
Linear Formula:
H2N(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
179.29
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing