Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3,4-Dihydro-2
H
-pyran-2-methanol
Synonym(s):
2-Hydroxymethyl-3,4-dihydro-2
H
-pyran, 3,4-Dihydro-2
H
-pyran-2-ylmethanol
Empirical Formula (Hill Notation):
C
6
H
10
O
2
CAS Number:
3749-36-8
Molecular Weight:
114.14
EC Number:
223-150-0
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Puriss (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
55665
3,4-Dihydro-2
H
-pyran-2-methanol,
≥98.5% (GC)
Expand
View Pricing