Skip to Content
Merck
2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride pharmaceutical impurity standard

2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride

2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride pharmaceutical impurity standard
Synonym(s):
2-[(4-Methyl-α-phenylbenzyl)oxy]ethyl(dimethyl)ammonium chloride, 4-Methyldiphenhydramine hydrochloride, N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamin hydrochloride, Neobenodine
Empirical Formula (Hill Notation):
C18H23NO · HCl
CAS Number:
Molecular Weight:
305.84
EC Number:

Products

Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type